Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

205532-M

Millipore

C₁₂E₈, PROTEIN GRADE® Detergent, 10% Solution

A non-ionic detergent useful for the solubilization of membrane-bound proteins in their native state. Aggregation number: 123; HLB number: 13.1. Note: 1 set = 5 x 10 ml.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H58O9
CAS Number:
Molecular Weight:
538.75
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

aggregation number

123

impurities

≤20 μM Peroxide

CMC

110 μM

storage temp.

2-8°C

SMILES string

O(CCOCCOCCOCCOCCCCCCCCCCCC)CCOCCOCCOCCO

InChI

1S/C28H58O9/c1-2-3-4-5-6-7-8-9-10-11-13-30-15-17-32-19-21-34-23-25-36-27-28-37-26-24-35-22-20-33-18-16-31-14-12-29/h29H,2-28H2,1H3

InChI key

YYELLDKEOUKVIQ-UHFFFAOYSA-N

General description

A non-ionic detergent useful for the solubilization of membrane-bound proteins in their native state. CMC = 110 µM; Aggregation number: 123; HLB number = 13.1.
Note: 1 set = 5 x 10 ml.

Warning

Toxicity: Irritant (B)

Physical form

10% aqueous solution.

Other Notes

Noirclerc-Savoye, M., et al. 2002. Prot. Expr. Purif.30, 18.
Jimenez, M., et al. 2001. Lipids36, 1169.
Casey, J.R., and Reithmeier, R.A. 1993. Biochemistry32, 1172.
Andersen, J.P., et al. 1983. Eur. J. Biochem.134, 205.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service