Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203890-M

Millipore

CRAC Channel Inhibitor, BTP2

The CRAC Channel Inhibitor, BTP2, also referenced under CAS 223499-30-7, controls the biological activity of CRAC Channel. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

CRAC Channel Inhibitor, BTP2, [N-{4-[3,5-bis(Trifluoromethyl)-1H-pyrazol-1-yl]phenyl}-4-methyl-1,2,3-thiadiazole-5-carboxamide], TPRC3 Channel Inhibitor II, Pyr2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9F6N5OS
CAS Number:
Molecular Weight:
421.32
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1[n](nc(c1)C(F)(F)F)c2ccc(cc2)NC(=O)c3[s]nnc3C

InChI

1S/C15H9F6N5OS/c1-7-12(28-25-23-7)13(27)22-8-2-4-9(5-3-8)26-11(15(19,20)21)6-10(24-26)14(16,17)18/h2-6H,1H3,(H,22,27)

InChI key

XPRZIORDEVHURQ-UHFFFAOYSA-N

General description

A cell-permeable pyrazole that acts as a more potent blocker of Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 590 nM by Fura-2; peak current density pA/pF = -1.13 and -7.50, respectively, with or without 3 µM BTP2/Pyr2 by whole cell clamp) than the transient receptor potential cation channel TRPC3-mediated ROCE (receptor-operated Ca2+ entry) inTRPC3-expressing HEK293 cells (IC50 = 4.21 µM by Fura-2; pA/pF = -1.54 and -18.50, respectively, with or without 3 µM BTP2/Pyr2). A great complement to Pyr 3 (Cat. No. 648490), Pyr6, and Pyr10 in Ca2+ signaling studies.

Biochem/physiol Actions

Primary Target
CRAC Channel
Target IC50: 590 nM by Fura-2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Schleifier, H., et al. 2012. Br J PharmacolIn Press
Yonetoku, Y., et al. 2006. Bioorg. Med. Chem.14, 5370.
Yonetoku, Y., et al. 2006. Bioorg. Med. Chem.14, 4750.
Zitt, C., et al. 2004. J. Biol. Chem.279, 12927.
Trevillyan, J.M., et al. 2001. J. Biol. Chem.276, 48118.
Djuric, S.W., et al. 2000. J. Med. Chem.43, 2975.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service