Skip to Content
MilliporeSigma

203783

5-Bromo-4-chloro-3-indolyl-β-D-glucuronic Acid, Cyclohexylammonium Salt

≥98% (HPLC), solid, β-glucuronidase substrate, Calbiochem®

Synonym(s):

5-Bromo-4-chloro-3-indolyl-β-D-glucuronic Acid, Cyclohexylammonium Salt, X-GlcA, X-Gluc, BCIG, BCI-β-D-glucuronide, BCI-glucuro, X-Glucuro, CHA-X-Gluc

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H26BrClN2O7
CAS Number:
Molecular Weight:
521.79
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352106
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

5-Bromo-4-chloro-3-indolyl-β-D-glucuronic Acid, Cyclohexylammonium Salt, Substrate for β-glucuronidase that produces insoluble, intense indigo-blue chromophore (615 nm) after enzymatic hydrolysis.

Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

soluble (2% in DMF)

shipped in

ambient

storage temp.

−20°C

General description

Substrate for β-glucuronidase that produces insoluble, intense indigo-blue chromophore (615 nm) after enzymatic hydrolysis. Useful for a variety of applications, including use in bacterial detection systems (e.g. E. coli).

Other Notes

Jefferson, R.A., et al. 1986. Proc. Natl. Acad. Sci. USA83, 8447.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library