Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.77
Form:
solid
Solubility:
water: soluble
Product Name
Protease Inhibitor Cocktail I, The Protease Inhibitor Cocktail I controls the activity of Protease. This small molecule/inhibitor is primarily used for Biochemicals applications.
Quality Segment
form
solid
manufacturer/tradename
Upstate®
solubility
water: soluble
shipped in
dry ice
Application
Use generally to inhibit serine, cysteine, aspartic, and metalloproteases
Physical form
2mM AEBSF, 1mM EDTA, 130μM Bestatin, E-64 14μM , 1μM Leupeptin, 0.3μM Aprotinin when reconstituted in 100ml of buffer; supplied as lyophilized powder
Preparation Note
5 years at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Protease Inhibitor Cocktail I
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20-201 | 08436037123948 |
