Skip to Content
MilliporeSigma

20-158

0.5M EDTA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

manufacturer/tradename

Upstate®

shipped in

wet ice

General description

Ethylenediaminetetraacetic acid (EDTA) is an aminopolycarboxylic acid (APCAs). It serves as a metal chelating agent. EDTA is an anticoagulant, which prevents blood clotting by chelating calcium from the blood. EDTA-mediated platelet aggregation leads to pseudothrombocytopenia.. It is used to treat poisoning caused by heavy metals such as mercury and lead.

Application

0.5M EDTA has been used as a constituent of Dulbecco′s phosphate‐buffered saline (DPBS) for resuspension of trypsinized Human embryonic kidney 293T (HEK293T) cells in ataxin‐1 aggregation kinetics assay. It may also be used in chromatin immunoprecipitation (ChIP) assays.

Physical form

0.5M EDTA, pH 8.0.

Preparation Note

1 year at 4°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


signalword

Warning

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

target_organs

Respiratory Tract

pictograms

Health hazardExclamation mark

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 Inhalation



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library