Skip to Content
MilliporeSigma

194430

ABTS Chromophore, Diammonium Salt

Synonym(s):

ABTS Chromophore, Diammonium Salt, 2,2ʹ-Azino-bis(3-ethylbenzthiazoline-6-sulfonic Acid), 2NH4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H18N4O6S4 · (NH3)2
CAS Number:
Molecular Weight:
548.68
UNSPSC Code:
41116133
NACRES:
NA.41
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Synthetic chemical)

Quality Segment

description

RTECS - DL7002000

assay

≥98% (HPLC)

form

crystals, powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)c1cc2c(cc1)N(\C(=N\N=C3/Sc4c(ccc(c4)[S](=O)(=O)O)N/3CC)\S2)CC.N.N

InChI

1S/C18H18N4O6S4.2H3N/c1-3-21-13-7-5-11(31(23,24)25)9-15(13)29-17(21)19-20-18-22(4-2)14-8-6-12(32(26,27)28)10-16(14)30-18;;/h5-10H,3-4H2,1-2H3,(H,23,24,25)(H,26,27,28);2*1H3/b19-17-,20-18-;;

InChI key

OHDRQQURAXLVGJ-AXMZSLBLSA-N

General description

Useful as a substrate for ELISA; acts as a hydrogen donor in peroxidase reactions. Radical cation has absorption maximum at 414 nm.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 1 week at 4°C or for up to 4 months at -20°C.

Other Notes

Groome, N.P. 1980. J. Clin. Chem. Clin. Biochem.18, 345.
Childs, R.E. and Bardsley, W.G. 1975. Biochem. J.145, 93.

Legal Information

ABTS is a trademark of Boehringer Mannheim Corp.
ABTS is a trademark of Roche
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library