Skip to Content
MilliporeSigma

1.93643

Sodium acetate trihydrate

greener alternative

Greener Alternative

EMPLURA®

Synonym(s):

Sodium acetate trihydrate, Acetic acid sodium salt trihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
UNSPSC Code:
12352302
Beilstein/REAXYS Number:
3732037
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

EMPLURA®

Quality Segment

autoignition temp.

1112 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

room temp

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives. Sodium acetate trihydrate supports sustainable technologies as an eco-friendly phase change material for thermal energy storage, offering efficient heat retention and release while reducing energy waste and promoting cleaner energy solutions.Click here for more information.

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Sodium acetate trihydrate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library