Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93607

Supelco

Sodium hypochlorite solution

~4 % (w/w) (available chlorine)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302

grade

laboratory grade

product line

EMPLURA®

form

solution

concentration

~4 % (w/w) (available chlorine)

pH

12-13 (20 °C, 68 °F)

bp

102 °C/1,013  hPa

mp

-20 °C (-4 °F)

solubility

water: soluble

density

1.22 g/cm3 at 20 °C (68 °F)

application(s)

industrial hygiene

storage temp.

20-25°C

SMILES string

[O-]Cl.[Na+]

InChI

1S/ClHO.Na/c1-2;/h2H;

InChI key

KQPBSBAEBKRAAU-UHFFFAOYSA-N

Related Categories

Packaging

1.93607.1021: 1 L Minimum Order Quantity: 8
1.93607.5021: 5 L Minimum Order Quantity: 4
1.93607.9026: 25 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service