Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.93000

Supelco

Sulfuric acid about 98%

for analysis EMPARTA® ACS

Synonym(s):

Sulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352322

grade

ACS reagent

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

product line

EMPLURA®

form

liquid

concentration

≤100%

pH

0.3 (25 °C, 49 g/L, (77 °F))

bp

~290 °C (lit.)

mp

-20 °C ( -4 °F)

solubility

water: soluble

density

1.840 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93000.0521: 500ML, Minimum order quantity: 16
1.93000.2521: 2.5L, Minimum order quantity: 4
1.93000.5021: 5 L Minimum Order Quantity: 2

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany
EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service