Skip to Content
MilliporeSigma

178440

Ferritin, Apo-, Equine Spleen

Ferritin, Apo-, Equine Spleen, CAS 9013-31-4, is a native protein shell of ferritin molecule lacking iron.

Synonym(s):

Ferritin, Apo-, Equine Spleen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25
MDL number:
Form:
lyophilized solid
Assay:
≥90% (SDS-PAGE)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - CD9365000

assay

≥90% (SDS-PAGE)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

dilute buffer: 1 mg/mL, water: 1 mg/mL

cation traces

Fe: <0.01%

shipped in

wet ice

storage temp.

−20°C

General description

Native apo-ferritin from equine spleen. Protein shell of ferritin molecule lacking iron. Large amounts are present in β-cells of the pancreas where it acts as an antioxidant.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

McDonald, M.J., et al. 1994. FASEB J. 8, 777.
de Silva, D., et al. 1992. Arch. Biochem. Biophys.298, 259.
Sun, S., et al. 1992. J. Biol. Chem.267, 25160.
Crichton, R.R. 1974. Structure Bonding17, 67.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library