Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

17-630

Sigma-Aldrich

ChIPAb+ Acetyl-Histone H4 - ChIP Validated Antibody and Primer Set

serum, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.52

biological source

rabbit

Quality Level

antibody form

serum

clone

polyclonal

species reactivity

eukaryotes, human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
cell based assay: suitable
immunoprecipitation (IP): suitable
multiplexing: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H4 set includes purified rabbit polyclonal antiserum and the normal rabbit IgG antiserum, which can be used to demonstrate that the acetyl-histone H4 antibody is capable of precipitating acetyl-histone H4 associated chromatin. The qPCR primers included flank the human GAPDH promoter and produce a 166 base pair PCR product.

Specificity

Predicted broad cross-reactivity among eukaryotes based upon sequence similarity.
Recognizes acetylated histone H4 of approximately 10 kDa. Cross-reacts with acetylated histone H2B from Tetrahymena and weakly crossreacts with acetylated histone H2B from HeLa cells.
May cross-react with other acetylated proteins.

Immunogen

The acetyl-histone H4 antiserum is made against a peptide corresponding to amino acids 2-19 of Tetrahymena histone H4.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This ChIPAb+ Acetyl-Histone H4 -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.
Western Blot Analysis:
Acidextracted proteins from normal HeLa cells (Lane 1) and HeLa cells treated with 5 mM sodium butyrate for 24 hours (Lane 2) were resolved by electrophoresis, transferred to nitrocellulose and probed with anti-acetyl Histone H4 (1:2000).
Proteins were visualized using a goat-anti rabbit secondary antibody conjugated to HRP and a chemi-luminescence detection system. (Please see figures).

Packaging

25 assays per set. ~4 μL per chromatin immunoprecipitation

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from untreated HeLa cells (1 X 106 cell equivalents) was subjected to chromatin immunoprecipitation using 4 μL of either a normal rabbit antiserum or Anti-Acetyl Histone
H4 serum and the Magna ChIP A (Cat. # 17-610) Kit. Successful immunoprecipitation of acetyl histone H4 associated DNA fragments was verified by qPCR using control ChIP Primers flanking the human GAPDH promoter (Please see figures).
Please refer to the EZ-Magna A ChIP (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

10 kDa

Physical form

Anti-Acetyl-Histone H4 (rabbit polyclonal IgG). One vial containing 100 μL of antiserum containing 0.05% sodium azide. Store at -20°C.

Normal Rabbit Serum. One vial containing 100 uL antiserum containing 0.05% sodium azide. Store at -20°C.

Control Primers. One vial containing 75 μL of 5 μM of each primer specific for for human GAPDH.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Control
Included negative control rabbit IgG antiserum and control primers specific for human GAPDH promoter.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunjian Huang et al.
Journal of immunology (Baltimore, Md. : 1950), 190(9), 4470-4473 (2013-04-02)
Regulatory T cells (Tregs) play a pivotal role in the maintenance of immunological self-tolerance. Deficiency or dysfunction of Tregs leads to severe autoimmune diseases. Although the forkhead/winged-helix family member FOXP3 is critical for Treg differentiation and function, the molecular basis
Rosita Accardi et al.
Carcinogenesis, 36(11), 1440-1451 (2015-10-02)
Although Epstein-Barr virus (EBV) infection is widely distributed, certain EBV-driven malignancies are geographically restricted. EBV-associated Burkitt's lymphoma (eBL) is endemic in children living in sub-Saharan Africa. This population is heavily exposed to food contaminated with the mycotoxin aflatoxin B1 (AFB1).
Rosita Accardi et al.
PLoS pathogens, 9(3), e1003186-e1003186 (2013-03-22)
Many studies have proved that oncogenic viruses develop redundant mechanisms to alter the functions of the tumor suppressor p53. Here we show that Epstein-Barr virus (EBV), via the oncoprotein LMP-1, induces the expression of ΔNp73α, a strong antagonist of p53.
Qiyuan Kuai et al.
Journal of medical virology, 90(9), 1478-1485 (2018-04-29)
Highly active antiretroviral therapy can reduce the human immunodeficiency virus (HIV) viral load in the plasma to undetectable levels. However, because of the presence of latent HIV reservoirs, it is difficult to completely eradicate HIV in infected patients. Our objective
Sachith Mettananda et al.
Nature communications, 8(1), 424-424 (2017-09-06)
β-Thalassemia is one of the most common inherited anemias, with no effective cure for most patients. The pathophysiology reflects an imbalance between α- and β-globin chains with an excess of free α-globin chains causing ineffective erythropoiesis and hemolysis. When α-thalassemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service