Skip to Content
MilliporeSigma
All Photos(1)

Documents

171601

Sigma-Aldrich

Amyloid Precursor Protein β-Secretase Inhibitor

The Amyloid Precursor Protein β-Secretase Inhibitor controls the biological activity of Amyloid Precursor Protein β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

Amyloid Precursor Protein β-Secretase Inhibitor, KTEETSEVN(stat)VAEF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H118N16O27
Molecular Weight:
1651.81
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

aqueous buffer, pH 7.5: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent inhibitor of the amyloid precursor protein (APP) β-secretase (IC50 = 30 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
APP
Product does not compete with ATP.
Reversible: no
Target IC50: 30 nM against amyloid precursor protein (APP) β-secretase

Warning

Toxicity: Standard Handling (A)

Sequence

H-Lys-Thr-Glu-Glu-Ile-Ser-Glu-Val-Asn-Stat-Val-Ala-Glu-Phe-OH [Stat = (3S,4S)-Statine]

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sinha, S., et al. 1999. Nature 402, 537.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Hao Wang et al.
Progress in neurobiology, 202, 102032-102032 (2021-03-16)
ApoE4, an apolipoprotein implicated in cholesterol transport and amyloid-β (Aβ) metabolism, is a major genetic risk determinant for Alzheimer's Disease (AD) and drives its pathogenesis via Aβ-dependent and -independent pathways. C/EBPβ, a proinflammatory cytokines-activated transcription factor, is upregulated in AD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service