Skip to Content
MilliporeSigma

Skip To

1.70337

Niobium ICP standard

traceable to SRM from NIST NH4NbF6 in H2O 1000 mg/l Nb Certipur®

Synonym(s):

Niobium Calibration Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

product line

Certipur®

concentration

1000 mg/L Nb in water

technique(s)

ICP: suitable

pH

3 (20 °C in H2O)

density

1.000 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Niobium ICP standard is a suitable calibration standard for elemental analysis by Inductively Coupled Plasma (ICP) technique.

Analysis Note

1000 mg/l Nb in water, prepared with high-purity, NH4NbF6 and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.703361.703131.70341
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

technique(s)

ICP: suitable

technique(s)

ICP: suitable

technique(s)

ICP: suitable

technique(s)

ICP: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

application(s)

industrial qc
pharmaceutical

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

Certipur®

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters