Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.70317

Supelco

Europium ICP standard

traceable to SRM from NIST Eu₂O₃ in HNO₃ 2-3% 1000 mg/l Eu Certipur®

Synonym(s):

Europium Calibration Solution for ICP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

product line

Certipur®

concentration

1000 mg/L Eu in 2-3% HNO3

technique(s)

ICP: suitable

pH

0.5 (20 °C in H2O)

density

1.012 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Europium ICP standard is a suitable calibration standard for elemental analysis by Inductively Coupled Plasma (ICP) technique.

It can serve as calibration standard for Inductively Coupled Plasma Mass Spectrometry (ICP-MS) during the chemical analysis of luminescent metal hydride samples doped with Eu.

Analysis Note

1000 mg/L Eu in 2-3% nitric acid, prepared with high-purity Eu2O3 and HNO3

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recovery rate and homogeneity of doping europium into luminescent metal hydrides by chemical analysis
R Hahn
Royal Society of Chemistry Advances, 5, 9722-9726 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service