Skip to Content
MilliporeSigma

1.70303

Arsenic ICP standard

traceable to SRM from NIST H3AsO4 in HNO3 2-3% 1000 mg/l As Certipur®

Synonym(s):

Arsenic Calibration Standard for ICP/AAS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

suitable for EPA 200.7, suitable for EPA 200.8

product line

Certipur®

concentration

1000 mg/L As in 2-3% HNO3 ( )

technique(s)

ICP: suitable

pH

0.5 (20 °C in H2O)

density

1.013 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Arsenic ICP standard can be used as a calibration standard in the determination of trace elements in soy sauce by inductively coupled plasma-mass spectrometry (ICP-MS).

Analysis Note

1000 mg/L As in 2-3% nitric acid, prepared with high-purity H3AsO4, HNO3, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library