Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.70207

Supelco

Beryllium standard solution

Be₄O(C₂H₃O₂)₆ in HNO₃ 0.5 mol/l 1000 mg/l Be Certipur®

Synonym(s):

Beryllium Calibration Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

suitable for EPA 200.7
suitable for EPA 200.8

product line

Certipur®

technique(s)

AAS: suitable

pH

0.5 (20 °C in H2O)

density

1.017 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Berylliumstandard solution may be used as a calibration standard in the determination of beryllium in urine by atomic absorption spectrometry.

Analysis Note

1000 mg/L Be in 0.5 mol/L HNO3, prepared with high-purityBe4O(C2H3O2)6, HNO3, and water

Other Notes

For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Skin,Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beryllium and its inorganic compounds-Determination of beryllium in urine by atomic absorption spectrometry [Biomonitoring Methods, 2017]
Paul R, et al.
The MAK-Collection for Occupational Health and Safety, 2, 1690-1709 (2002)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service