Skip to Content
MilliporeSigma

16-219

Anti-c-Myc Tag Antibody

UPSTATE®, monoclonal, 4A6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Clone:
4A6, monoclonal
Species reactivity:
human
Application:
IP
Citations:
18
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Myc Tag Antibody, clone 4A6, agarose conjugate, clone 4A6, Upstate®, from mouse

biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

Epitope tags are short peptide sequences that are easily recognized by tag-specific antibodies. Due to their small size, epitope tags do not affect the tagged protein’s biochemical properties. Most often sequences encoding the epitope tag are included with target DNA at the time of cloning to produce fusion proteins containing the epitope tag sequence. This allows anti-epitope tag antibodies to serve as universal detection reagents for any tag containing protein produced by recombinant means. This means that anti-epitope tag antibodies are a useful alternative to generating specific antibodies to identify, immunoprecipitate or immunoaffinity purify a recombinant protein. The anti-epitope tag antibody is usually functional in a variety of antibody-dependent experimental procedures. Expression vectors producing epitope tag fusion proteins are available for a variety of host expression systems including bacteria, yeast, insect and mammalian cells.
Varies

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc.

Application

Anti-Myc Tag Antibody, clone 4A6, agarose conjugate detects level of Myc Tag & has been published & validated for use in IP.
Immunoaffinity Purification:
A previous lot was used to purify Myc-tagged proteins. Elute with 100 mM tetraethyl ammonium (TEA), pH 11.5.

Biochem/physiol Actions

Reactivity with other species has not been confirmed.

Features and Benefits

Format: Gel Immobilized

Physical form

Purified mouse monoclonal IgG1 covalently coupled to protein G agarose beads and provided as a 50% slurry in buffer containing PBS, pH 7.4, and 0.05% sodium azide. Liquid suspension.

Preparation Note

Stable for 1 year at 2-8ºC from date of receipt. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Cells transfected with Myc-tagged fusion vector.
Routinely evaluated by immunoprecipitation of Myc-tagged Akt/PKB from a COS transfected cell lysate, subsequent detection using anti-Myc Tag (05-724).

Immunoprecipitation Analysis:
4 μg of anti-Myc Tag, clone 4A6, agarose conjugate immunoprecipitated Myc-tagged Akt/PKB from 250 μg of a COS transfected cell lysate, which was then detected by immunoblot analysis, using 0.5 μg/mL of anti-Myc Tag, clone 4A6 (Catalog # 05-724).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library