Skip to Content
MilliporeSigma
All Photos(1)

Documents

16-212

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 4A6, biotin conjugate

clone 4A6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

Immunogen: peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc

Specificity: Recognizes and is specific for recombinant proteins containing the Myc epitope tag (EQKLISEEDL) in a variety of sequence contexts. Also recognizes human Myc.

Species Cross-reactivity: Human, others not tested

Ig Type: IgG

Host: Mouse

Molecular Weight: Varies depending upon the protein which is tagged

Application

Detect Myc Tag using this Anti-Myc Tag Antibody, clone 4A6, biotin conjugate validated for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

routinely evaluated by western blot on Myc-tagged recombinant protein

Physical form

PBS and 0.05% sodium azide before the addition of glycerol to 30%

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Zhen Liu et al.
CNS neuroscience & therapeutics, 24(5), 394-403 (2018-01-25)
Neural tube defects (NTDs) are birth defects of the nervous system and are the second most frequent cause of birth defects worldwide. The etiology of NTDs is complicated and involves both genetic and environmental factors. CASP9 is an initiator caspase
Jian Sun et al.
Cell reports, 38(5), 110312-110312 (2022-02-03)
The Zic family of zinc finger transcription factors plays a critical role in multiple developmental processes. Using loss-of-function studies, we find that Zic5 is important for the differentiation of retinal pigmented epithelium (RPE) and the rod photoreceptor layer through suppressing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service