Skip to Content
MilliporeSigma

1.59166

Acetic acid solution

greener alternative

30%, for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Acetic acid solution, Methane carboxylic acid solution, Methylformic acid solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
NACRES:
NA.21
UNSPSC Code:
12352106
MDL number:
Beilstein/REAXYS Number:
506007
Concentration:
29.0-31.0 % (w/v) (acetic acid)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetic acid 30%, for analysis EMSURE® Reag. Ph Eur

agency

reag. Ph. Eur.

Quality Segment

product line

EMSURE®

form

liquid

expl. lim.

4-19.9 % (v/v) acetic acid)

greener alternative product characteristics

Waste Prevention
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

29.0-31.0 % (w/v) (acetic acid)

transition temp

flash point >100 °C

density

1.039 g/cm3 at 20 °C

greener alternative category

storage temp.

no temp limit

SMILES string

CC(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

General description

We are committed to providing greener alternatives that adhere to the 12 Principles of Greener Chemistry. This acetic acid is bio-renewable and produced from sustainably sourced beechwood, minimizing waste and promoting resource efficiency. Thus it aligns with ′Waste Prevention′, and ′Use of Renewable Feedstocks′.

Analysis Note

Assay: 29.0 - 31.0 % (w/v)

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Acetic acid 30%


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 1

flash_point_f

212.0 °F

flash_point_c

> 100 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
159166050004022536241326