Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.50413

Supelco

Purospher® STAR RP-18 endcapped (3 µm) HPLC Columns

L × I.D. 125 mm x 3 mm Hibar® RT HPLC Column

Synonym(s):

Purospher® STAR RP-18 endcapped (3µm) Hibar® RT 125-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Purospher® STAR RP-18 endcapped (3 µm), L × I.D. 125 mm x 3 mm Hibar® RT HPLC column

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Purospher®

feature

endcapped

packaging

1 ea of

extent of labeling

17% Carbon loading

parameter

400 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

12.5 cm × 3 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

C18 bonding phase

particle size

3 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

reversed phase
supercritical fluid (SFC)

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

Purospher® STAR RP-18 endcapped, 3 µm HPLC columns are designed for universal use and allow the separation of basic, neutral, and metal chelating compounds with simple mobile phases and excellent peak symmetry. They are based on ultra-pure fully porous silica particles (Type B). These columns offer an outstanding stability from pH 1.5 to 10.5 over a wide temperature range, and suitability for up to 100% aqueous mobile phases. Plus, they demonstrate best all-round retention characteristics, as proven by the Tanaka test. Thanks to these features, Purospher® STAR RP-18 endcapped, 3 µm HPLC columns allow maximum flexibility to use the best chromatographic conditions for any separation including reversed phase.Purospher® STAR RP-18 endcapped, 3 µm Hibar® RT 125-3 is a ready to use stainless steel column suitable for connection to any HPLC system.The column dimension of 125 mm length x 3 mm I.D. is the optimal choice for high resolution HPLC separation of complex samples at lower flow rate, allowing significant solvent savings and improved sensitivity. A 3 mm I.D. column enables a more sustainable separations in comparison to 4 or 4.6 mm I.D. column due to the use of less solvent for the chromatographic separation.The recommended guard column is Purospher® STAR RP-18 endcapped (5 µm) LiChroCART® 4-4 part number 1.50250.0001]. For direct connection to a Hibar® RT columns the Precolumn holder for 4-4 LiChroCART® cartridges, part number 1.51487.0001 can be used.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, amino acids, esters, fatty acids, ketones, nitrosamines, nucleosides, PCBs, peptides, phenols, preservatives, steroids, and metabolized steroids.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Best all-round performance according to Tanaka
  • Outstanding pH stability from pH 1.5 to 10.5
  • No phase collapse when using highly aqueous mobile phases
  • Good suitability for LC-MS applications

Analysis Note

Pressure: ≤ 120 bar
Theoretical Plates (N/m) (Anthracene): ≥ 130000
Symmetry (Anthracene): 0.8 - 1.4
Capacity factor (Anthracene): 4.0 - 6.0

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service