Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.50270

Supelco

Purospher®STAR RP-8 endcapped (5 µm) HPLC Columns

L × I.D. 4 mm x 4 mm LiChroCART® Guard-cartridge

Synonym(s):

Purospher® STAR RP-8 endcapped (5 µm) LiChroCART® 4-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Purospher® STAR RP-8 endcapped (5 µm), L × I.D. 4 mm x 4 mm LiChroCART® Guard-cartridge

material

stainless steel column

Quality Level

Agency

suitable for USP L7

product line

Purospher®

feature

endcapped

packaging

10 pak of

extent of labeling

11.2% Carbon loading

parameter

250 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

0.4 cm × 4 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

C8 bonding phase

particle size

5 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

reversed phase

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

Purospher® STAR RP-8 endcapped, 5 µm HPLC columns are designed for universal use and allow the separation of basic, neutral, and metal chelating compounds with simple mobile phases and excellent peak symmetry. They are based on ultra-pure fully porous silica particles (Type B). Purospher® STAR RP-8 endcapped, 5 µm LiChroCART® 4-4 is a guard cartridge which allows the integration into a manuCART cartridge column holder part number 1.51486.0001 for direct, dead volume free connection to a analytical LiChroCART® cartridge column with inner dimension or 2, 3, 4, or 4.6 mm I.D. and a column length of 75, 100, 125, 150 or 250 mm.For direct connection to a Hibar® RT columns the Precolumn holder for 4-4 LiChroCART® cartridges, part number 1.51487.0001 can be used.For connection to any other analytical HPLC column with capillary connection the Precolumn holder for 4-4 LiChroCART® cartridges, part number 1.16217.0001 is suitable.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, amino acids, esters, fatty acids, ketones, nitrosamines, nucleosides, PCBs, peptides, phenols, preservatives, steroids, and metabolized steroids.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Outstanding pH stability from pH 1.5 to 10.5
  • Good suitability for LC-MS applications

Analysis Note

Selectivity: passes test

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
MANUCART is a trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service