Skip to Content
MilliporeSigma
All Photos(1)

Documents

1468520006

Millipore

Fluid Thioglycollate Medium

suitable for sterility testing, pkg of 1 unit (1L)

Synonym(s):

FTM, FTM with 0.1% Tween®80, Fluid Thioglycollate Medium (FTM, Thioglycollate Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit (1L)

pH

6.9±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid Thioglycollate Medium (FTM) with 0.1% Tween®80 is a complex medium designed for the cultivation and isolation of fastidious aerobic and anaerobic microorganisms present in sterility test samples of pharmaceutical industry.
The medium formulation contains a reduced agar content thereby rendering the appearance to be clear. The anaerobic atmosphere is created through reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment.
Further reduction of oxygen diffusion is induced by agar, while maintaining the clarity of the medium. The indicator ingredient present in the medium indicates the reduction potential. Increase in oxygen concentration is indicated through a color change reaction in the fluid thioglycollate (FTM) medium.

Application

Fluid Thioglycollate Medium (FTM) with 0.1% Tween®80 is used as a sterility testing medium for sterile pharmaceutical samples. FTM clear is mainly used to cultivate and isolate anaerobic and microaerophilic microbes.

Analysis Note

The appearance of the medium is clear or almost clear and yellowish.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service