Skip to Content
MilliporeSigma

Skip To

1.46783

Tryptic Soy Agar - Ready-to-use Contact Plates

Tween®, Sodium Thiosulfate, Lecithin, Histidine, lid, ICR Plus: lockable, plate diam. 55 mm, irradiated, triple packed of 10 plates, suitable for surface monitoring

Synonym(s):

TSA, Soybean Casein Digest Agar, ICRplus contact plate, Tryptic Soy Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.74
UNSPSC Code:
41106231
Composition:
Histidine , Lecithin , Sodium Thiosulfate , Tween®
Plate diam.:
55 mm
Feature:
lid ICR Plus: lockable, ready-to-use
Sterility:
irradiated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene plate

Quality Segment

agency

EP, USP

description

ICR plus with lockable lid

sterility

irradiated

form

plate

feature

lid ICR Plus: lockable, ready-to-use

composition

Histidine , Lecithin , Sodium Thiosulfate , Tween®

packaging

triple packed of 10 plates

plate diam.

55 mm

application(s)

air monitoring
pharmaceutical, surface monitoring

compatibility

for use with MAS-100®

storage temp.

2-25°C

suitability

bacteria, fungi

General description

Tryptic Soy Contact Agar + LTHThio contact ICR+ is designed for the determination of the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Clean rooms and Isolators. It is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The formulation of the Soybean Casein-Digest medium contains combination of casein and soy peptones supplying organic nitrogen, particularly amino acids and longer-chained peptides. The medium is supplemented with neutralizers like. lecithin, Tween®, histidine, sodium thiosulfate and pyruvate to inactivate potential residues of disinfectants. In addition, ICRplus plates (plus or “+”) are supplied with a lockable lid for safe transport after sampling without the risk of losing the lid as well as for aerobic (“CLOSED”-position) and for anaerobic/ microaerophilic incubation in the “VENT” position.

Application

Tryptic Soy Agar (TSA, Soybean Casein Digest Agar) is a complex medium for cultivation and isolation of a wide range of bacteria, yeasts and molds. The media finds its application in environmental monitoring by determining the total microbial aerobic and anaerobic count on dry, sanitized surfaces and personnel in Isolators and Clean Rooms.

Legal Information

MAS-100 is a registered trademark of MBV AG
TWEEN is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.465521.461951.46231
plate diam.

55 mm

plate diam.

55 mm

plate diam.

55 mm

plate diam.

55 mm

suitability

bacteria, fungi

suitability

bacteria, fungi, molds, yeasts

suitability

bacteria, fungi

suitability

bacteria, fungi

application(s)

air monitoring
pharmaceutical, surface monitoring

application(s)

air monitoring, pharmaceutical

application(s)

pharmaceutical, surface monitoring

application(s)

microbiology
pharmaceutical, surface monitoring

packaging

triple packed of 10 plates

packaging

pkg of 10 plates Triple packed, box of 20 ea, box of 200 ea

packaging

pkg of 10 plates Triple packed, box of 20 ea, box of 200 ea

packaging

pkg of 10 plates Triple packed, box of 20 ea, box of 200 ea

sterility

irradiated

sterility

irradiated

sterility

sterile; γ-irradiated

sterility

irradiated

feature

lid ICR Plus: lockable, ready-to-use

feature

ready-to-use

feature

ready-to-use

feature

ready-to-use


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. salve, queste piastre corrispondono a petri-tadtm petri dishes oppure RTU1&2 plates o RTU 3-tolso? Devo comprare un audit che copre l'item in oggetto e non sono sicura. Grazie

    1 answer
    1. 146783 is part of RTU 1&2 production line.

      Helpful?

Reviews

No rating value

Active Filters