Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46691

Millipore

Fluid A

ready-to-use, bottle volume 800 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 septa
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable

bottle capacity

1000 mL

bottle volume

800 mL , filling volume

pH

7.1±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid A is a general rinse buffer designed as a clear colorless liquid in compliance with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06.
Each bottle has a screw cap with septum closure to reduce the risk of cross-contamination and optimize the disinfection procedures.

Application

Fluid A is used to rinse aqueous solutions during sterility testing by membrane filtration. It is also used to dilute soluble solids for the same application and as a rinsing fluid for membrane filtration of non-sterile products.
The product is robust and suitable for:
  • Dissolving or diluting samples,
  • Reconstituting commercial microorganism sample.
  • As a transport medium for microorganisms.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service