Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.46627

Millipore

Tryptic Soy Broth Ready-to-use Bottles

Lecithin , Tween® 80, bottle capacity 125 mL, bottle filling volume 90 mL, closure type, blue screw cap with 3 loci, box of 10 bottles

Synonym(s):

CASO Broth, Soyabean Casein Digest Broth, TSB, TSB with neutralizer, Tryptic Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225

Agency

EP
JP
USP

Quality Level

description

for the isolation and cultivation of fastidious aerobic bacteria, yeasts, & molds

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type blue screw cap with 3 loci
ready-to-use

composition

Lecithin
Tween® 80

packaging

box of 10 bottles

bottle capacity

125 mL

bottle filling volume

90 mL

pH

7.3 (25 °C)

application(s)

food and beverages
pharmaceutical
sterility testing

components

tween
lecithin

storage temp.

2-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth (TSB) with neutralizers is a universal complex and nutritious medium and supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi. Lecithin, along with Polysorbate (Tween®) 80, is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrols (PHB esters).

Application

Tryptic Soy Broth with LT is used for the isolation and cultivation of fastidious aerobic bacteria, yeasts, and molds in the presence of disinfectant residues. The medium is designed for the enumeration of microorganisms in sterile or non-sterile products from the food and pharmaceutical industry.

Features and Benefits

Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy and reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service