Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NC.81
UNSPSC Code:
41122409
Feature:
autoclavable
Material:
stainless steel rack (1.4301 el. polished)
material
stainless steel rack (1.4301 el. polished)
Quality Segment
feature
autoclavable
diam. × H
70 mm × 250 mm
application(s)
cosmetics
food and beverages
pharmaceutical
surface monitoring
compatibility
for use with 55mm Contact Plates
storage temp.
no temp limit
General description
Racks for ICR (Isolator Clean Room) and ICR plus contact plates are suitable for monitoring sanitized, dry surfaces in critical cleanrooms, RABS, and isolators. The stainless-steel racks aid in safe, easy, and convenient handling.
Application
Racks for ICR Contact Plates are used for safe and convenient handling of contact plates in critical cleanrooms, RABS, and isolators
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.