Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46477

Millipore

Fluid A

ready-to-use, bottle volume 700 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type crimp cap with septum
ready-to-use

packaging

bottle of

technique(s)

sterile filtration: suitable

bottle capacity

1000 mL

bottle volume

700 mL , filling volume

pH

7.0±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid A is ageneral rinse buffer designed in compliance with the European Pharmacopeia (EP)2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06.The product is a clear colorless liquid.
Each bottle has a crimpcap with septum closure to reduce the risk of cross-contamination and optimizethe disinfection procedures.

Application

Fluid A isused to rinse aqueous solutions during sterility testing by membrane filtration.It is also used to dilute soluble solids for the same application and as arinsing fluid for membrane filtration of non-sterile products.

The productis robust and suitable for:
Dissolving or dilutingsamples,
reconstituting commercialmicroorganism sample.
As a transport medium formicroorganisms.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service