Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.46220

Millipore

Thioglycollate Medium

box of 100 tubes, ready-to-use, tube volume 9 mL , filling volume

Synonym(s):

Fluid Thioglycollate Medium, Thioglycollate Broth, FTM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

description

For the isolation and cultivation of fastidious anaerobic and aerobic microorganisms

sterility

sterile; autoclaved

form

liquid (ready-to-use)

shelf life

6 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

box of 100 tubes
single packed of (9 ml in 17 ml tube with red cap)

tube capacity

17 mL

tube volume

9 mL , filling volume

pH

7.1

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid Thioglycolate Medium is a universal complex medium. It is recommended for sterility testing of pharmaceutical products according to the European and US Pharmacopoeia. Thioglycolate and L-Cystine in the medium reduce the redox potential of the culture medium to create an anaerobic atmosphere. Also, mercury and other heavy metal compounds are inactivated by these agents. The content of agar further reduces a rapid diffusion of oxygen through the medium but may lead to slight turbidity in larger volumes (filled bottles). Resazurin indicates the reduction potential of the medium. An increased concentration of oxygen is indicated by a color change to pink.

Application

Fluid Thioglycolate Medium is used for the isolation and cultivation of fastidious anaerobic as well as for aerobic microorganisms. This is used for sterility control of pharmaceutical products.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service