Skip to Content
MilliporeSigma

1.46166

Rogosa Agar

with TMB+Peroxidase, plate volume 18 mL, pack of 100 plates, selective for Lactobacillus

Synonym(s):

Lactobacillus Agar, Lactobacillus Selective Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352201
Composition:
with TMB+Peroxidase
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterilization compatibility

autoclavable compatible

Quality Segment

composition

with TMB+Peroxidase

packaging

pack of 100 plates

technique(s)

microbiological culture: suitable

plate volume

18 mL

pH

5.5±0.2 (25 °C)

application(s)

food and beverages

storage temp.

2-8°C

suitability

selective for Lactobacillus

General description

Rogosa Agar with TMB and peroxidase is a nutrient rich selective media for the isolation and enumeration of lactobacilli spp. The accompanying microbial flora is inhibited by the high acetate concentration and low pH of the medium.
Tween® 80 provides necessary fatty acids for metabolism during growth. Lower concentrations of manganese, magnesium and iron ensure optimal growth of lactobacillus present in the test samples from food industry.

Application

Rogosa Agar with TMB and peroxidase is used to isolate and enumerate lactobacilli spp from test samples from food industry.

Analysis Note

Poured plates are clear and yellowish brown.

Legal Information

TWEEN is a registered trademark of Croda International PLC


Still not finding the right product?

Explore all of our products under Rogosa Agar


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library