Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46141

Millipore

Buffered NaCl-Peptone Solution

ready-to-use, tube volume 9.9 mL , Filling Volume

Synonym(s):

Buffered Saline Peptone Water, Buffered Sodium Chloride Peptone Water, NaCl Peptone Solution (Buffered)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106229
NACRES:
NA.74

Agency

EP 2.6.13

Quality Level

sterility

autoclaved

form

liquid

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

tube of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

tube capacity

17 mL

tube volume

9.9 mL , Filling Volume

pH

7.0

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical

storage temp.

2-25°C

General description

Buffered NaCl-Peptone Solution has low peptone content, along with phosphate buffer and sodium chloride to maintain the viability of microorganisms present without supporting growth.

Application

Buffered NaCl-Peptone Solution is designed for the microbial examination of non-sterile pharmaceutical products. It can also be used as a solution for sample preparation, dilution, or washing.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service