Skip to Content
MilliporeSigma

14-531-M

EGFR Protein, active, 10 µg

Active, N-terminal GST-tagged human EGFR kinase domain, amino acids 696-end, for use in Enzyme Assays.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
eCl@ss:
32160405
Biological source:
human
Mol wt:
Mw 86 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

mol wt

Mw 86 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

UniProt accession no.

General description

N-terminal GST-tagged human EGFR kinase domain, amino acids 696-end
Product Source: Human EGFR expressed by baculovirus in Sf21 insect cells.

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-531M when ordering the 250 µg size.

Analysis Note

Routinely evaluated by phosphorylation of Angiotensin II peptide substrate

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library