Skip to Content
MilliporeSigma

14-510-M

Lyn Protein, active, 10 µg

Active, N-terminal his6-tagged recombinant full-length human Lyn, for use in Kinase Assays.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
eCl@ss:
32160405
Biological source:
human
Recombinant:
expressed in Sf21 cells
Mol wt:
Mw 59.6 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in Sf21 cells

mol wt

Mw 59.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal his6-tagged recombinant full-length human Lyn
Product Source: expressed in Sf21 cells

Application

Kinase Assays: Depending upon the specific activity of the lot , 7.6 -15.3ng of enzyme phosphorylated 0.1 mg/ml poly (Glu, Tyr) 4:1 {Sigma P7244} in a standard kinase assay. See technical datasheet included with purchased product for lot specific activity values.
Research Category
Metabolism
Research Sub Category
Obesity

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Lyn
Target Sub-Family: TK

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-510M when ordering the 250 µg size.

Physical form

Ni2+/NTA-agarose

Preparation Note

6 months at -70°C

Analysis Note

routinely evaluated by phosphorylation of Poly (Glu4-Tyr) (4:1)

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library