Skip to Content
MilliporeSigma

1.44154

PyroDetect Kit

Reagents and Interleukin-1 ELISAβ kit for Monocyte Activation test (MAT), for Pyrogen testing

Synonym(s):

Pyrogen testing kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116129
Agency:
EP Chapter 2.6.30
Suitability:
pyrogenic bacteria
Manufacturer/tradename:
PyroDetect
Application(s):
pharmaceutical
pyrogen testing
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

EP Chapter 2.6.30

Quality Segment

feature

ready-to-use

packaging

pkg of 1 kit

manufacturer/tradename

PyroDetect

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect Systems

shipped in

ambient

storage temp.

2-8°C

suitability

pyrogenic bacteria

General description

PyroDetect Kit is a Monocyte Activation Test (MAT) for the detection of pyrogens. This test allows the determination of the pyrogenic activity in a solution or suspension by inducing interleukin production in cell cultures with human monocytes.

Application

The PyroDetect Kit is used for the detection and quantification of the pyrogenic activity in pharmaceutical products to ensure their safety.

Biochem/physiol Actions

The PyroDetect System uses cryopreserved human whole blood as a source of monocytes. The response to pyrogenic substances is determined by measurement of the Interleukin-1β (IL-1β) production. The ELISA (enzyme-linked immunosorbent assay) microplate in the PyroDetect System is precoated with an antibody specific for IL-1β. Interleukin molecules present in the culture supernatant, transferred from the blood cultures, are bound to the immobilized antibody.
The unbound substances are washed away, and an enzyme-linked polyclonal antibody specific for IL-1β is added. The IL-1β bound complex is then detected in a color reaction initiated by the addition of a suitable substrate. The efficiency of IL-1β production in the pyrogen reaction is calculated from the data of a photometrical quantification of the colored product by the enzymatic reaction in an ELISA reader.

Features and Benefits

  • Ready-to-use kit
  • Greater product safety
  • Highly compatible
  • In vitro test with cryoblood
  • Reliable results
  • Regulatory recognition

Other Notes

Each PyroDetect Kit contains:
  • Cell Culture Microplate for 96 microcultures, with lid - 1 plate
  • Cell Culture Medium - RPMI with L-Glutamine and HEPES - 50 mL
  • IL-1β Microplate - 12 strips of 8 wells each, coated with a monoclonal antibody specific for IL-1β - 1 plate
  • IL-1β Conjugate
  • Wash Buffer Concentrate
  • Color Reagent Α
  • Color Reagent Β
  • Stop Solution
PyroDetect Cryoblood (1.44155)
Reference Standard Endotoxin (1.44161)
NEP Control of HKSA (MATHKSA)
NEP Control of Flagellin (MATFLAGELLIN)


pictograms

CorrosionExclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1 - Skin Sens. 1

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library