Skip to Content
MilliporeSigma

1.42020

Cyclodextrin HPB

EMPROVE® EXPERT, Ph. Eur., NF

Synonym(s):

Cyclodextrin HPB, (2-Hydroxypropyl)-β-Cyclodextrin, HP-B-CD, Hydroxypropyl Betadex

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C42H70-nO35 · (C3H7O)n
CAS Number:
UNSPSC Code:
12352201
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

NF, Ph. Eur.

product line

EMPROVE® EXPERT

form

solid

mol wt

avg mol wt 1387.2 g/mol

pH

5.0-7.0 (20 g/L in H2O)

solubility

450 g/L

application(s)

liquid formulation
ophthalmics
pharmaceutical
solid formulation
solubility enhancement

storage temp.

15-25°C

General description

When working on a pharmaceutical formulation, there are specific considerations to make and challenging hurdles to take before you can successfully launch your final drug product - formulation stability, release kinetics and bioavailability limitations just to name a few. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Cyclodextrin HPB (hydroxypylated β-cyclodextrin) can be used to strongly increase solubility of active pharmaceutical ingredients (APIs) by forming API:cyclodextrin inclusion complexes. It can also increase API stability and safety as well as mask undesired odors and tastes. The broad application range of Cyclodextrin HPB comprises liquid, semi-solid and solid formulations, as well as diverse routes of administration. Due to its low microbial and endotoxin limits, Cyclodextrin HPB Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_c

> 400 °C

flash_point_f

>752.0 °F



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library