Skip to Content
MilliporeSigma

1.40183

Modified UVM Listeria selective enrichment broth

EcoCult®, according to USDA-FSIS, powder, for Listeria spp., pkg of 5 kg, pkg of 10 kg

Synonym(s):

UVM Modified Listeria Enrichment Broth, Modified University of Vermont Medium, UVM, UVM1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213
Manufacturer/tradename:
EcoCult®
Agency:
according to USDA-FSIS
Form:
powder
Suitability:
Listeria spp. (spp.)
Application(s):
food and beverages
microbiology
pathogen testing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

according to USDA-FSIS

Quality Segment

form

powder

packaging

pkg of 10 kg, pkg of 5 kg

manufacturer/tradename

EcoCult®

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

10-30°C

suitability

Listeria spp. (spp.)

General description

EcoCult® Modified UVM Listeria selective enrichment broth has a combination of various peptones, extracts, salts, and buffer substances, which enable the growth of Listeria spp. The selectivity is due to the antiproliferative substances nalidixic acid and acriflavine hydrochloride. The two-stage enrichment method has demonstrated its value, especially with sample materials (meat and meat products) that are characterized by a high level of accompanying flora.

Application

EcoCult® Modified UVM Listeria selective enrichment broth is suitable for the selective enrichment of Listeria monocytogenes from red meat, poultry, ready-to-eat Siluriformes (fish) and egg products, and environmental samples.

Features and Benefits

  • Compliance to major regulatory standards ISO, FDA-BAM, EPA or USP
  • Clear labeling and Certificate of Analysis to check compliance at a glance
  • Reliable and accurate quality in powder format
  • Cost-effective pricing

Legal Information

ECOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library