Skip to Content
MilliporeSigma

137672

SyntheChol® Synthetic Cholesterol

For Cell Culture, EMPROVE® EXPERT

Synonym(s):

Cholesterol, 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
UNSPSC Code:
12352212
Beilstein/REAXYS Number:
1915888
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

product line

EMPROVE® EXPERT

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

application(s)

pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

General description

SyntheChol® synthetic cholesterol, for Cell Culture Emprove® Expert, is a synthetic cholesterol supplement that has been developed to specifically meet the demands of the life science and biopharmaceutical industries. Due to the synthetic nature of the material, the elimination of animal-derived components reduces the incidence of performance variability in the medium and eliminates safety risks due to adventitious agents associated with these components. With a newly optimized manufacturing process, this product is available in quantities from grams to tons to support your development progress.

Application

This supplement is designed to be added to our chemically defined,animal component-free CHO and insect (Sf-RVN) cell media, as well as HEK-293media for VGT manufacturing. This material has been tested for compatibilitywith CHO and Sf-9 cell lines. Cholesterol provides structure to cellularmembranes, stores energy, and is vital in cell signaling and in regulating cellfunctions.
This product is intended for research or further manufacturing but notfor human or therapeutic use.

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Packaging

137672.0001 /1g /
137672.0010 / 10 g /
137672.0100 / 100 g /
137672.1000 / 1000g /

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under SyntheChol® Synthetic Cholesterol


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library