Skip to Content
MilliporeSigma

137151

2-Mercaptoethanol

EMPROVE® EVOLVE

Synonym(s):

β-Mercaptoethanol, 2-Hydroxyethylmercaptan, BME, Thioethylene glycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HSCH2CH2OH
CAS Number:
Molecular Weight:
78.13
UNSPSC Code:
12161504
Beilstein/REAXYS Number:
773648
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

2.69 (vs air)

Quality Level

vapor pressure

1 mmHg ( 20 °C)

form

liquid

expl. lim.

18 %

concentration

14.3 M (pure liquid), 99.0-101.0 % (w/w)

impurities

≤1.0% related substances (GC) 2,2-Dithiodiethanol

refractive index

n20/D 1.500 (lit.)

bp

157 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

suitability

corresponds for identity (IR)

application(s)

pharma/biopharma processes

storage temp.

2-8°C

SMILES string

OCCS

InChI

1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2

InChI key

DGVVWUTYPXICAM-UHFFFAOYSA-N

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Application

BME is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for SDS-PAGE at a concentration of 5%. Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on SDS-PAGE. Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure.

Analysis Note

purity (GC) ≥99.0%

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 2-Mercaptoethanol


signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_c

74 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library