Skip to Content
MilliporeSigma

1.37134

Acetonitrile

EMPROVE® ESSENTIAL

Synonym(s):

Acetonitrile, Methyl cyanide, Anhydrous ACN, Dry MeCN

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
UNSPSC Code:
12191502
Beilstein/REAXYS Number:
741857
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

assay

≥99.5% (GC)

form

liquid (colorless)

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-25°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

General description

We offer a range of high quality critical raw materials, solvents and reagents plus the documentation and expertise to meet stringent regulatory requirements. In addition to our comprehensive portfolio, we also offer custom manufacturing services to meet your exact needs. As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Acetonitrile is relatively inert, due to the strong C-C bond which can only be hydrolyzed under extreme conditions, making it an excellent reaction solvent used in pharma production as a reagent, in the reaction or extraction solvent, starting material to synthesize vitamins A and B1, cortisone, carbonate drugs and some amino acids. Acetonitrile is also used as a solvent in the production of insulin and antibiotics.
Acetonitrile Emprove® Essential is suitable for pharmaceutical application.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Acetonitrile


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library