Skip to Content
MilliporeSigma

1.37048

SAFC

D-(+)-Glucose

anhydrous EMPROVE® EXPERT Ph Eur,BP,USP,ACS

Pharma Manufacturing

Synonym(s):

Dextrose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724615
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-075-1

grade

ACS reagent

Quality Level

Agency

BP
Ph. Eur.
USP

product line

EMPROVE® EXPERT

form

solid

autoignition temp.

500 °C

potency

25800 mg/kg LD50, oral (Rat)

mp

150-152 °C (lit.)

solubility

470 g/L

bulk density

630 kg/m3

application(s)

parenterals

storage temp.

15-25°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Analysis Note

Assay (calc. on anhydrous substance): 97.5 - 102.0 %
Identity (IR): passes test
Identity (spec. rotation calc. on anhydrous substance): passes test
Identity (HPLC): passes test
Identity (water): passes test
Appearance of solution: passes test
Color and clarity of solution: passes test
Titrable acid: ≤ 0.002 meq/g
Conductivity: ≤ 20 µS/cm
Spec. rotation (α25/D): 52.5 - 53.0 °
In water insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
Related substances [sum of impurities A (maltose) and B (isomaltose)!: ≤ 0.4 %
Related substances [Impurity C (maltotriose)!: ≤ 0.2 %
Related substances [Impurity D (fructose)!: ≤ 0.15 %
Related substances (unspecified impurities): ≤ 0.10 %
Related substances (Sum of all impurities): ≤ 0.5 %
Dextrin: passes test
Soluble starch, sulfite: ≤ 15 ppm
Starch: passes test
Residual solvents (ICH Q3C): excluded by the manufacturing process
Loss on drying (105 °C): ≤ 0.2 %
Water (according to Karl Fischer): ≤ 1.0 %
Sulfated ash (600 °C): ≤ 0.02 %
Microbiological purity(Total aerobic microbial count (TAMC)): ≤ 100 CFU/g
Microbiological purity(Total yeast and mould count (TYMC)): ≤ 100 CFU/g
Bile tolerant Gram negative Bacteria(absent in 1 g): passes test
Escherichia coli (absent in 1 g): passes test
Pseudomonas aeruginosa (absent in 1 g): passes test
Staphylococcus aureus (absent in 1 g): passes test
Candida albicans (absent in 1 g): passes test
Salmonella ssp (absent in 10 g): passes test
Endotoxins: ≤ 10 I.U./g
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Corresponds to Ph Eur,BP,USP,ACS

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service