Skip to Content
MilliporeSigma

1.32383

Anaerocult C

For creating a low-oxygen, high-CO2 atmosphere inside an anaerobic jar, suitable for microbiology

Synonym(s):

Microaerophilic growth, Microaerophilic incubation

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41104308
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

feature

Generates an oxygen depleted and CO2-enriched atmosphere in a 2.5 liter anaerobe jar.

packaging

pack of 25 bags

manufacturer/tradename

Anaerocult

storage condition

(Store dry and tightly closed.

Keep in a well-ventilated place.)

application(s)

microbiology

compatibility

ANAEROTEST®

storage temp.

15-25°C

General description

The Anaerocult C system is used specifically for culturing Campylobacter species and other microorganisms with fastidious requirements. The special formulation of the reagent mixture is designed so that the cultivation jar reaches gas concentrations of 8-10% vol. CO2 and 5-6% vol. O2.

Application

Anaerocult C is used to generate an oxygen-depleted and CO2-enriched atmosphere in a 2.5 L anaerobe jar. For the application, the inoculated Petri dishes and the moistened Anaerocult C bag are placed in the anaerobic jar which is closed subsequently for further culturing.

Legal Information

ANAEROCULT is a trademark of Merck KGaA, Darmstadt, Germany
ANAEROTEST is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation markHealth hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Lungs, Respiratory system

Storage Class

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library