Skip to Content
MilliporeSigma

12660

Albumin, Bovine Serum, Fraction V, Modified Cohn, pH 5.2

Synonym(s):

Albumin, Bovine Serum, Fraction V, Modified Cohn, pH 5.2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.25
UNSPSC Code:
12352202
eCl@ss:
32160409
Assay:
≥96% (SDS-PAGE)
Form:
solid
Impurities:
≤5% Moisture
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Merck USA index - 14, 8468

Quality Level

assay

≥96% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤5% Moisture

color

off-white

solubility

0.15 M sodium chloride: 10 mg/mL

cation traces

heavy metals: ≤50 ppm

shipped in

ambient

storage temp.

2-8°C

General description

Albumin, Bovine Serum, Fraction V, Modified Cohn, pH 5.2 is a native fatty acid profile that reflects endogenous lipids. It has a pH in the range of 4.8-5.4 in 150 mM NaCl (1%).
Bovine serum albumin (BSA), a non-glycosylated globular, α-helical protein, belongs to the serum albumin family. It consists of three domains with two sub-domains each and has 17-disulfide bonds.

Application

Albumin, Bovine Serum, Fraction V, Modified Cohn, pH 5.2 has been used as a blocking agent in western blotting. It has also been used as a component in phosphate-buffered saline (PBS) for diluting secondary antibodies and blocking in immunofluorescence to observe the localization of the Golgi apparatus.

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is the most vital component of cell culture media. It aids in the differentiation of embryonic stem cells (hESC). It plays a key role in the transportation of drugs, hormones, and fatty acids. BSA is used in enzyme-linked immunosorbent assay (ELISA) as a blocking agent.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Anna Eslami et al.
Journal of visualized experiments : JoVE, (44) (2010-12-30)
Western blotting is an analytical technique used to detect specific proteins in a given sample of tissue homogenate or extract. It uses gel electrophoresis to separate native or denatured proteins by the length of the polypeptide (denaturing conditions) or by
Miyu Tsuchiya et al.
Oncology letters, 11(5), 3279-3286 (2016-04-29)
N-glycosylation is a post-translational protein modification with a wide variety of functions. It has been predicted that R-spondin1 (RSPO1) is N-glycosylated, although this remains unknown. The present study identified that RSPO1 was N-glycosylated at Asn137, and that N-glycosylation of RSPO1
Tamara Topală et al.
Clujul medical (1957), 87(4), 215-219 (2014-01-01)
The continuous search for new molecules with therapeutic abilities has led to the synthesis and characterization of a large number of metal complexes, proven to exhibit potential as pharmacological agents through their antibacterial, antiviral, antifungal and antineoplastic properties. As serum



Global Trade Item Number

SKUGTIN
A8097-10ML04061833391204
12660-1KG04055977222630
12660-100GM04055977222647