Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.25193

SAFC

Cellvento® 4HEK Liquid

Chemically defined, sterile; sterile-filtered, suitable (for HEK293 cell culture)

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

description

shipped with cooling pack

sterility

sterile; sterile-filtered

product line

Cellvento®

form

liquid

technique(s)

cell culture | mammalian: suitable (suspension)

storage temp.

2-8°C

General description

Cellvento® 4HEK Medium is a chemically defined medium optimized for adeno-associated virus (AAV) production in multiple human embryonic kidney (HEK) 293 lineages. Cellvento® 4HEK Medium has been streamlined and cell culture media components incorporated to increase media stability and reduce protein oxidation. The formulation is animal component free.

Application

Cellvento® 4HEK Medium can be used as amplification and production medium for gene therapy applications. This medium enables low doubling time and high viable cell densities with multiple HEK293 cell lines. Cellvento®4HEK Medium is compatible with commonly used transfection reagents like PEI, supports AAV production and facilitates high titers of full capsids in various HEK293 lineages. This easy-to-use medium can be used for all stages like growth, transfection, and production.

Quantity

Formulated to contain 30.8 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Read our white paper on the development of a novel cell culture medium for AAV production with multiple HEK293 lineages and process optimization.

Read our white paper on the development of a novel cell culture medium for AAV production with multiple HEK293 lineages and process optimization.

Read our white paper on the development of a novel cell culture medium for AAV production with multiple HEK293 lineages and process optimization.

Read our white paper on the development of a novel cell culture medium for AAV production with multiple HEK293 lineages and process optimization.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service