Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.24823

Sigma-Aldrich

Nitro blue tetrazolium chloride (NBT)

for microscopy

Synonym(s):

Nitro blue tetrazolium chloride (NBT), 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis [2-(4-nitrophenyl)-5-phe nyl-2H-tetrazolium chloride], NBT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
817.64
MDL number:
UNSPSC Code:
41116124
EC Index Number:
206-067-4

Quality Level

form

solid

mp

189 °C

εmax

800-900 at 256-259 nm in methanol at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nitro blue tetrazolium chloride (NBT) is a dry dye for the staining of histological tissue sections and histochemical as well as in vivo methods on material of human origin.
The dye is a CE certified IVD product and has a constant dye content and a reliable high quality, which is one of the major precondition for a reproducible diagnostic staining. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Dry dye used in histochemistry

Analysis Note

Identity (UV/VIS-Spectrum)passes testAbsorption maximum λ max (Methanol)256 - 259 nmSpec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; methanol; calc. on dried substance)800 - 900Loss on drying (Vacuum < 7 mbar, P₂O₅, 24 h)≤ 12 %Suitability for microscopypasses test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service