Skip to Content
MilliporeSigma

1.24802

5-Aminolevulinic acid hydrochloride

for biochemistry

Synonym(s):

5-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochlorid, 5-Aminolevulinic Acid Hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H9NO3 · xHCl
CAS Number:
Molecular Weight:
131.13 (free base basis)
UNSPSC Code:
12352107
EC Index Number:
226-679-5
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

solid

mp

150-156 °C

solubility

100 g/L

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3]CC(=O)CCC(=O)O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H

InChI key

ZLHFONARZHCSET-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99.0 %
Identity (IR-spectrum): conforms
Melting range: 150 - 156 °C
Heavy metals (as Pb): ≤ 0.001 %
TLC-Test: conforms


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
124802902504022536520841
124802250004022536220079
124802050004022536220062