Skip to Content
MilliporeSigma

124017

Akt Inhibitor VIII, Akti-1/2

InSolution, ≥95%, Isozyme-Selective

Synonym(s):

InSolution Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C34H29N7O
Molecular Weight:
551.64
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
liquid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

wet ice

storage temp.

−70°C

General description

A cell-permeable, potent, and selective inhibitor of Akt1/Akt2 activity (IC50 = 58 nM, 210 nM, and 2.12 µM for Akt1, Akt2, and Akt3, respectively, in in vitro kinase assays). The inhibition is reported to be pleckstrin homology (PH) domain-dependent. It does not exhibit any inhibitory effect against PH domain-lacking Akts, or other closely related AGC family kinases, PKA, PKC, and SGK, even at concentrations as high as 50 µM. Overcomes Akt1/Akt2-mediated resistance to chemotherapeutics in tumor cells and is shown to block basal and stimulated phosphorylation/activation of Akt1/Akt2 both in cultured cells in vitro and in mice in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Akt1, Akt2, Akt3
Product does not compete with ATP.
Reversible: yes
Target IC50: 58 nM, 210 nM, and 2.12 µM for Akt1, Akt2, and Akt3, respectively, in in vitro kinase assays

Packaging

Packaged under inert gas
Please refer to vial label for lot-specific concentration.

Physical form

A 10 mM (1 mg/181 µl) solution of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018) in DMSO.

Preparation Note

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Barnett, S.F., et al. 2005. Biochem. J.385, 399.
DeFeo-Jones, D., et al. 2005. Mol. Cancer Ther.4, 271.
Zhao, Z., et al. 2005. Bioorg. Med. Chem. Lett.15, 905.
Lindsley, C.W., et al. 2005. Bioorg. Med. Chem. Lett.15, 761.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Enrico Desideri et al.
Autophagy, 19(1), 152-162 (2022-04-19)
Impaired degradation of the transcriptional coactivator YAP1 and IL6ST (interleukin 6 cytokine family signal transducer), two proteins deregulated in liver cancer, has been shown to promote tumor growth. Here, we demonstrate that YAP1 and IL6ST are novel substrates of chaperone-mediated
Sarah J Russell et al.
The Journal of biological chemistry, 295(45), 15292-15306 (2020-09-02)
Muscle atrophy is regulated by the balance between protein degradation and synthesis. FOXO1, a transcription factor, helps to determine this balance by activating pro-atrophic gene transcription when present in muscle fiber nuclei. Foxo1 nuclear efflux is promoted by AKT-mediated Foxo1



Global Trade Item Number

SKUGTIN
124017-1MG07790788047993