Skip to Content
MilliporeSigma

124012

Akt Inhibitor V, Triciribine

Synonym(s):

InSolution Akt Inhibitor V, Triciribine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H16N6O4
CAS Number:
Molecular Weight:
320.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - RY8455000

Quality Segment

form

solid

solubility

soluble (DMSO (10 mg/ml). Further dilute at aqueous buffer in low micromolar concentrations.)

storage temp.

2-8°C

SMILES string

[n]2(c3ncnc4c3c(c2)C(=NN4C)N)[C@@H]1O[C@@H]([C@H]([C@H]1O)O)CO

InChI

1S/C13H16N6O4/c1-18-11-7-5(10(14)17-18)2-19(12(7)16-4-15-11)13-9(22)8(21)6(3-20)23-13/h2,4,6,8-9,13,20-22H,3H2,1H3,(H2,14,17)/t6-,8-,9-,13-/m1/s1

InChI key

HOGVTUZUJGHKPL-HTVVRFAVSA-N

General description

A cell-permeable and reversible tricyclic nucleoside that selectively inhibits the cellular phosphorylation/activation of Akt1/2/3 by targeting an Akt effector molecule other than PI 3-K or PDK. Exhibits little effect towards cellular signaling pathways mediated by PKC, PKA, SGK, Stat3, p38, ERK1/2, or JNK. Shown to preferentially induce apoptosis and growth arrest in cancer cells with aberrant Akt activity both in vitro (≥60% in cell proliferation at 20 µM) and in vivo (≥80% inhibition in tumor growth in mice at 1 mg/kg/day, i.p.). Also available in InSolution format (Cat. No. 124038.


Still not finding the right product?

Explore all of our products under Akt Inhibitor V, Triciribine


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library