Skip to Content
MilliporeSigma

12-360

Acetyl-Histone H3 Peptide

Lys9/14

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetyl-Histone H3 (Lys9/14) Peptide, Routinely evaluated by Dot Blot Assay. This peptide was spotted on nitrocellulose & detected by using polyclonal anti-acetylated Histone H3 (06-599).

Quality Segment

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... H3F3B(3021)

Physical form

Lyophilized powder

Preparation Note

Lyophilized: 2 years at 4°C; Rehydrated: 6 months at -20°C.

Analysis Note

Routinely evaluated by Dot Blot Assay. This peptide was spotted on nitrocellulose and detected by using polyclonal anti-acetylated Histone H3 (06-599).

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library