Skip to Content
MilliporeSigma

12-359

Acetyl-Histone H3 Peptide

Lys14

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetyl-Histone H3 (Lys14) Peptide, Routinely evaluated by Dot Blot Assay. This peptide was recognized on a dot blot using Anti-acetyl Histone H3 (Lys14). The peptide was not recognized by anti-acetyl Histone H3 (Lys9) or anti-acetyl Histone H3 (Lys9,14).

Quality Segment

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... H3F3B(3021)

Physical form

Lyophilized powder

Preparation Note

2 years at -20°C

Analysis Note

Routinely evaluated by Dot Blot Assay. This peptide was recognized on a dot blot using Anti-acetyl Histone H3 (Lys14), (06-911). The peptide was not recognized by anti-acetyl Histone H3 (Lys9) or anti-acetyl Histone H3 (Lys9,14).

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Acetyl-Histone H3 (Lys14) Peptide


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library