Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41105330
NACRES:
NA.41
eCl@ss:
32011807
Technique(s):
western blot: suitable
Species reactivity:
-
Application:
WB
Citations:
7
biological source
human
Quality Segment
form
liquid
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
shipped in
dry ice
Biochem/physiol Actions
A431
Physical form
modified RIPA buffer (50mM Tris-HCl, pH 7.4, 1% NP40, 0.25% sodium deoxycholate, 150mM NaCl, 1mM EDTA, 1mM PMSF, 1μg/ml each of aprotinin, leupeptin, pepstatin, 1mM Na3VO4, 1mM NaF) and diluted with non-reducing sample buffer (31mM Tris-HCl, pH 6.8, 5% glycerol, 1% SDS, 0.002% bromphenol blue).
Preparation Note
1 year at -20°C
Analysis Note
Routinely evaluated as a positive control for several of Upstates antibodies.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1 - Eye Irrit. 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
