Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-218

Sigma-Aldrich

Tyrosine Phosphopeptide

lyophilized, TSTEPQpYQPGENL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.26

product name

Tyrosine Phosphopeptide (TSTEPQpYQPGENL),

manufacturer/tradename

Upstate®

Quality Level

technique(s)

activity assay: suitable

shipped in

wet ice

Biochem/physiol Actions

Protein Target: PTP-1B

Quality

Routinely evaluated using the phosphopeptide as a substrate for PTP-1B (14-358) in conjunction with a non-radioactive malachite green phosphate detection system, PTP Assay Kit 2 (17-126).

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylated synthetic peptides as tools for studying protein phosphatases.
L A Pinna et al.
Biochimica et biophysica acta, 1222(3), 415-431 (1994-07-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service